missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG/Nepmucin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
407.00€
Spécification
| Antigène | CD300LG/Nepmucin |
|---|---|
| Dilution | ELISA |
| Classification | Polyclonal |
| Conjugué | Janelia Fluor 549 |
| Forme | Purified |
Description
CD300LG/Nepmucin Polyclonal antibody specifically detects CD300LG/Nepmucin in Rat samples. It is validated for ELISASpécification
| CD300LG/Nepmucin | |
| Polyclonal | |
| Purified | |
| RUO | |
| CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
| Produced in rabbits immunized with purified, recombinant Rat CD300LG/Nepmucin (Accession#: XP_002724611.1; Met1-Arg160). | |
| Primary |
| ELISA | |
| Janelia Fluor 549 | |
| Rabbit | |
| 50mM Sodium Borate | |
| 146894 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit