Learn More
CD31/PECAM-1 Antibody (MEC13.3) - BSA Free, Novus Biologicals™
Code produit. 18758133 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.025 mL
0.1 mL
Conditionnement:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
18758133 0.025 mL 25µL
18737993 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit. 18758133 Fournisseur Novus Biologicals Code fournisseur NB6001475SS
Rat Monoclonal Antibody has been used in 17 publications
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.
TRUSTED_SUSTAINABILITY
Spécification
Antigène CD31/PECAM-1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay, CyTOF
Classification Monoclonal
Clone MEC13.3
Concentration 1.0 mg/mL
Conjugué Unconjugated
Dilution Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:100, In vitro assay reported in scientific literature (PMID 24647208), In vivo assay reported in scientific literature, Flow (Cell Surface) 2.5 ug/ml, CyTOF-ready
Alias de gène adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, MEC 13.3 CD31, MEC 13.3 Clone, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Symboles de gène(s) PECAM1
Espèces hôtes Rat
Immunogène This CD31/PECAM-1 Antibody (MEC13.3) was developed against mouse endothelial cell line T-end.
Poids moléculaire de l’antigène 82.5 kDa
Méthode de purification Protein G purified
Quantité 0.025 mL
État réglementaire RUO
Disciplines de recherche Angiogenesis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 5175
Espèces cibles Human, Mouse
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG2a κ
Afficher plus Afficher moins
For Research Use Only