missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio CD3e Mouse, Clone: C3e/1308, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 916-MSM8-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD3e | |
Cell surface and cytoplasmic | |
C3e/1308 | |
Unconjugated | |
NP_000724 | |
Mouse | |
20kDa | |
Protein A/G Purified | |
RUO | |
916 | |
IgG2b κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD3E | |
Recombinant human CD3e fragment (aa 23-119) | |
Jurkat cells. Tonsil and lymph node | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |