missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, Brilliant Ultraviolet 496, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 612937
Informations supplémentaires : Poids : 0.00020kg
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Spécification
CD4 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
SK3 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Peripheral Blood T Cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |