Learn More
CD4 Mouse, FITC, Clone: RPA-T4, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB12-F-25Tests
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15911518
  • 82.15€ / 25 tests
Expédition estimée: 12-06-2024
Spécification
CD4
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
Protein A/G Purified
RUO
920
IgG1 κ
Flow Cytometry
RPA-T4
FITC
NP_001181945
Mouse
25 Tests
Primary
Store at 2-4°C.