Learn More
CD45.1 Mouse, FITC, Clone: A20, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB42-F-500ug
Code nomenclature Nacres: NA.46
Code produit. 15992008
  • 288.75€ / 500µg
Spécification
CD45.1
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
A20
FITC
NP_001255215
Mouse
500 μg
Primary
Store at 2-4°C.