missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Mouse anti-Human, BUV737, Clone: HI100, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 612846
Informations supplémentaires : Poids : 0.00060kg
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry.Spécification
CD45RA | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
100 Tests | |
Primary | |
Human | |
IgG2b κ |
Flow Cytometry | |
HI100 | |
Mouse | |
RUO | |
CD45RA | |
Refrigerate (2 to 8°C) |