missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Mouse anti-Human, BB700, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566401
Informations supplémentaires : Poids : 0.01000kg
Description
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Flow Cytometry.Spécification
CD56 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
B159 | |
Mouse | |
RUO | |
Human | |
Purified |