Learn More
CD59 Monoclonal Antibody (MEM-43), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MHCD5901
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry.Spécification
CD59 | |
Monoclonal | |
FITC | |
CD59 | |
16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cb59b molecule; CD59; CD59 antigen; CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344); CD59 glycoprotein; CD59 molecule; CD59 molecule (CD59 blood group); CD59 molecule, complement regulatory protein; Cd59a; CD59a antigen; CD59A glycoprotein; Cd59b; CD59b antigen; CD59B glycoprotein; CD59b moleucle, complement regulatory protein; complement inhibitor CD59; complement regulatory protein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; human leukocyte antigen MIC11; Ly-6-like protein; lymphocytic antigen CD59/MEM43; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; membrane attack complex (MAC) inhibition factor; membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin; surface anitgen recognized by monoclonal antibody 16.3A5; T cell-activating protein | |
Mouse | |
purified | |
ASR | |
966 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
MEM-43 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P13987 | |
CD59 | |
Human CD59 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |