Learn More
CD62L / L-Selectin Rat, APC-Cy7, Clone: MEL-14, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB49-APC7-25ug
Code nomenclature Nacres: NA.46
Code produit. 15992108
  • 196.35€ / 25µg
Spécification
CD62L / L-Selectin
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Sell
Protein A/G Purified
RUO
20343
IgG2a κ
Flow Cytometry
MEL-14
APC-Cyanine7
NP_001157531
Rat
25 μg
Primary
Store at 2-4°C.