missing translation for 'onlineSavingsMsg'
Learn More

CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), Super Bright™ 702, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Affymetrix eBioscience 67-0079-41

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15923704

  • 163.00€ / 25 pièces
Expédition estimée: 15-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Description: The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. Applications Reported: This 53-6.7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 53-6.7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry.
Spécification

Spécification

CD7
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P09564
CD7
Affinity chromatography
RUO
924
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
eBio124-1D1 (124-1D1)
Super Bright 702
CD7
Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions