Learn More
CD71 / Transferrin Receptor Mouse, Clone: TFRC/1817, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 7037-MSM7-P0
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD71 / Transferrin Receptor | |
Cell surface | |
TFRC/1817 | |
Unconjugated | |
NP_001300894 | |
Mouse | |
85-95kDa (monomer) ;190kDa (dimer) | |
Protein A/G Purified | |
RUO | |
7037 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TFRC | |
Recombinant human TFRC protein fragment (aa94-212) | |
PC3 cells. Human placenta or breast carcinoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |