missing translation for 'onlineSavingsMsg'
Learn More

CD73 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: AD2, BD Horizon™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 612812

Informations supplémentaires : Poids : 0.00060kg

 Afficher plus de versions de ce produit

Code produit. 17257904

  • 590.00€ / 100 tests
Expédition estimée: 14-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The AD2 monoclonal antibody specifically binds to ecto-5’-nucleotidase, a 70 kDa, glycosyl phosphatidylinositol (GPI)-anchored glycoprotein. CD73 is expressed on subsets of T and B lymphocytes, follicular dendritic cells, epithelial cells, endothelial cells and mesenchymal stem cells. Its expression on lymphocytes increases during T and B cell development. CD73 has enzymatic activity and catalyzes the dephosphorylation of adenosine monophosphate (AMP) converting it to adenosine. It has been suggested that CD73 can mediate costimulatory signals in T cell activation and adhesion of lymphocytes to endothelium.

The antibody was conjugated to BD Horizon BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter).

Spécification

Spécification

CD73
Monoclonal
Brilliant Ultraviolet 737
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Flow Cytometry
AD2
Aqueous buffered solution containing ≤0.09% sodium azide.
Pre-B leukemia cell line
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions