Learn More
EnquireBio CD86 Mouse, PE, Clone: IT2.2, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB94-PE-100Tests
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD86 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD86 | |
Protein A/G Purified | |
RUO | |
942 | |
IgG2b κ |
Flow Cytometry | |
IT2.2 | |
PE | |
NP_787058 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |