Learn More
CD86 Rat anti-Mouse, FITC, Clone: GL1, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 553691
Informations supplémentaires : Poids : 0.01000kg
Description
The GL1 antibody has been reported to react with the B7-2 (CD86) costimulatory molecule expressed on a broad spectrum of leukocytes, including B lymphocytes, T lymphocytes, thioglycollate-induced peritoneal macrophages, dendritic cells and astrocytes. CD86 is expressed at low levels by freshly explanted peripheral B and T cells, and its expression is substantially increased by a variety of T cell- and B cell-specific stimuli with a peak expression after 18-42 hours of culture. In contrast to most naive CD4+ T cells, memory CD4+ T cells express B7-2, both at the mRNA and protein level. CD86, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. It has been shown to be involved in immunoglobulin class-switching and triggering of mouse NK cell-mediated cytotoxicity. CD80 (B7-1) is an alternate ligand for CD28 and CD152 (CTLA-4). GL1 antibody reportedly blocks MLR and stimulation of T cells by natural antigen-presenting cells. In addition, a mixture of anti-B7-1 and anti B7-2 (GL1) mAbs reportedly inhibits the in vitro interaction of CTLA-4 with its ligand and the in vivo priming of cytotoxic T lymphocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: GL1
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse (CBA/Ca) LPS-activated splenic B Cells
Flow Cytometry
Spécification
CD86 | |
Monoclonal | |
0.5mg/mL | |
B7-2; Ly-58; Cd28l2; Early T-cell costimulatory molecule 1; ETC1; MB7; CLS1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
GL1 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse (CBA/Ca) LPS-activated splenic B Cells | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.