missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90.2 (Thy-1.2) Monoclonal Antibody (30-H12), Super Bright™ 780, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
129.00€ - 379.00€
Spécification
| Antigène | CD90.2 (Thy-1.2) |
|---|---|
| Clone | 30-H12 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16361894
|
Invitrogen
78-0903-80 |
25 μg | |||||||
16371894
![]() |
Invitrogen™
78-0903-82 |
100 μg |
379.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Rat Monoclonal Antibody
CD90.2 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90.2 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90.2 is an 18 kDa plasma membrane antigen expressed by hematopoietic cells and neurons in all species studied, highly expressed in connective tissue, various fibroblast and stromal cell lines and lastly, by tumor endothelial cell lines. In the mouse, CD90.2 is expressed on all thymocytes and peripheral T lymphocytes.Specifications
| CD90.2 (Thy-1.2) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 21838 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| 30-H12 | |
| Flow Cytometry | |
| Super Bright 780 | |
| Rat | |
| Mouse | |
| P01831 | |
| Thy1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Thy1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title