Learn More
CD90 (Thy-1) Mouse, anti-Human, PerCP-Cyanine5.5, Clone: 5E10, STEMCELL Technologies
Mouse Monoclonal Antibody
Marque:  Stemcell Technologies 60045PS
Encompass_Preferred
Code nomenclature Nacres: NA.46
Code produit. 17178761
  • 722.25€ / 100 tests
Description
  • The 5E10 antibody reacts with CD90 (Thy-1), a GPI-linked membrane glycoprotein that is N-glycosylated at two sites, giving rise to 25 - 37 kDa molecules
  • CD90 has roles in signal transduction, cell adhesion and migration, neurite outgrowth, T cell activation, tumor suppression, and inhibition of the proliferation and differentiation of hematopoietic stem cells
  • It is a known ligand of β2 and β3 integrins and upregulates synthesis of fibronectin, osteonectin and thrombospondin
  • CD90 is broadly expressed, being found on human thymocytes, neurons, some glial cells, fibroblasts, activated endothelial cells, some leukemia cell lines and a distinct subset (<1%) of CD3+CD4+ T cells in human peripheral blood
  • This antibody clone has been verified for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411) and MesenCult™-XF Medium (Catalog #05420)
Spécification
Spécification
CD90 (Thy-1)
Monoclonal
PerCP-Cyanine5.5
100 Tests
Baboon, Cynomolgus, Rhesus Monkey, Pig, Human
Hematopoietic Stem and Progenitor Cells, Mammary Cells, Mesenchymal Stem and Progenitor Cells
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
5E10
Mouse
Primary
IgG1 κ
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.