missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Antibody (2Q42) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals NBP2-22000-0.025MG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Spezifikation
| CD97 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 1:50-1:250, CyTOF-ready | |
| AAH06676 | |
| CD97 | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein CD97. Rat monoclonal anti mouse CD97 AAH06676. | |
| 0.025 mg | |
| GPCR, Immunology | |
| 976 | |
| Mouse | |
| Purified |
| CyTOF, Flow Cytometry | |
| 2Q42 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur