missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD97 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13071-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
CD97 is a G-protein-coupled seven-span transmembrane adhesive receptor that is constitutively expressed on granulocytes and monocytes and rapidly upregulated on T and B cells upon activation. CD97 is produced in alternatively spliced forms and its cellular ligand is CD55 (DAF), which protects various cell types from complement-mediated damage. Interaction of CD97 on leukocytes and CD55 on vessel cells probably facilitate leukocyte activation and migration into the tissues, similarly, CD97 seems to play a role in tumour migration and invasiveness. CD97 is involved in T cell regulation and peripheral granulocyte homeostasis.Spécification
CD97 | |
Polyclonal | |
Unconjugated | |
CD97 | |
CD97, CD97 antigen, CD97 molecule, Leukocyte antigen CD97, TM7LN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
976 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48960 | |
CD97 | |
CD97 Fusion Protein Ag3745 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |