missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12087-2-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal domit polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.Spécification
CDA1 | |
Polyclonal | |
Unconjugated | |
TSPYL2 | |
CDA1, Cell division autoantigen 1, CINAP, CTCL, DENTT, NP79, Nuclear protein of 79 kDa, TSPX, TSPYL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64061 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H2G4 | |
TSPYL2 | |
CDA1 Fusion Protein Ag2722 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |