Learn More
CDC42SE2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA558472
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: EPTNFVHTAH VGSGDLFSGM NSVSSIQNQM QSKGGYGGGM PANVQMQLVD TKAG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Probably involved in the organization of the actin cytoskeleton by acting downstream of CDC42, inducing actin filament assembly. Alters CDC42-induced cell shape changes. In activated T-cells, may play a role in CDC42-mediated F-actin accumulation at the immunological synapse. May play a role in early contractile events in phagocytosis in macrophages.Spécification
CDC42SE2 | |
Polyclonal | |
Unconjugated | |
CDC42SE2 | |
2810404F18Rik; AA408783; AA536669; CDC42 small effector 2; CDC42 small effector protein 2; Cdc42se2; non-kinase Cdc42 effector protein SPEC2; RGD1563924; small effector of CDC42 protein 2; SPEC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRR3 | |
CDC42SE2 | |
Recombinant protein corresponding to Human CDC42SE2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only