Learn More
CDCP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA532703
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Panda, Horse (100%) Hamster, Rabbit (93%) Mouse, Rat, Dog, Elephant, Pig (87%).
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Spécification
CDCP1 | |
Polyclonal | |
Unconjugated | |
Cdcp1 | |
9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100065703, 64866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H5V8 | |
Cdcp1 | |
Synthetic 15 amino acid peptide from extracellular domain of human CDCP1. | |
50 μg | |
Primary | |
Equine, Human, Non-human Primate | |
Antibody | |
IgG |