missing translation for 'onlineSavingsMsg'
Learn More

CDK8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 22067-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831615

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CDK8 is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This kinase and its regulatory subunit cyclin C are components of the RNA polymerase II holoenzyme complex, which phosphorylates the carboxy-terminal domain (CTD) of the largest subunit of RNA polymerase II. This kinase has also been shown to regulate transcription by targeting the CDK7/cyclin H subunits of the general transcription initiation factor IIH (TFIIH), thus providing a link between the ′Mediator-like′ protein complexes and the basal transcription machinery.
Spécification

Spécification

CDK8
Polyclonal
Unconjugated
CDK8
CDK8, Cell division protein kinase 8, cyclin dependent kinase 8, K35, Mediator complex subunit CDK8, Protein kinase K35
Rabbit
Antigen Affinity Chromatography
RUO
1024, 264064
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49336, Q8R3L8
CDK8
CDK8 Fusion Protein Ag17148
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions