Learn More
CDR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA543665
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Peptide sequence: MLAENLVEEF EMKEDEPWYD HQDLQQDLQL AAELGKTLLD RNTELEDSVQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%
CDR2 Polyclonal antibody specifically detects CDR2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spécification
CDR2 | |
Polyclonal | |
Unconjugated | |
CDR2 | |
AA617262; CDR2; CDR62; cerebellar degeneration related protein 2; cerebellar degeneration-related 2; cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; major Yo paraneoplastic antigen; MGC128861 protein; paraneoplastic cerebellar degeneration-associated antigen; PCD17; RGD1310578; Yo; Yo paraneoplastic antigen | |
Rabbit | |
Protein A | |
RUO | |
1039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q01850 | |
CDR2 | |
Synthetic peptide directed towards the N-terminal of human CDR2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |