Learn More
CEACAM1 Monoclonal Antibody (283324), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA523908
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD31, rhICAM-1, -2, -3, recombinant mouse MAdCAM-1, or rhVCAM-1 was observed. In sandwich ELISAs, no cross-reactivity with rhCEACAM-3, rhCEACAM-5, or rhCEACAM-6 was observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CEACAM1 Monoclonal antibody specifically detects CEACAM1 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spécification
CEACAM1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P13688 | |
CEACAM1 | |
Mouse myeloma cell line NS0-derived recombinant human CEACAM-1 Gln35-Gly428 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
283324 | |
Unconjugated | |
CEACAM1 | |
antigen CD66; ATP-dependent taurocolate-carrier protein; bb-1; BGP; BGP1; BGP-1; BGPI; BGPR; Biliary glycoprotein 1; biliary glycoprotein D; bone gamma-carboxyglutamic acid (Gla) protein; carcinoembryonic antigen 1; carcinoembryonic antigen 7; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); Cc1; C-CAM; C-CAM 105; Ccam1; CD66a; CD66a antigen; Cea1; Cea-1; Cea7; Cea-7; CEACAM1; CEA-related cell adhesion molecule 1; CEA-related cell adhesion molecule 1 (bone gamma-carboxyglutamic acid (Gla) protein) (osteocalcin); Cell-CAM 105; ecto-ATPase; GP110; hepatitis virus (MHV-4) susceptibility; hepatitis virus receptor; Hv2; Hv-2; mCEA1; Mhv-1; MHVR; MHV-R; MHVR1; mmCGM1; mmCGM1a; mmCGM2; murine hepatitis virus receptor; osteocalcin; pp120; pp120/ecto-ATPase | |
Mouse | |
Protein A/G | |
RUO | |
634 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |