Learn More
CEACAM8/CD66b Antibody (24H12), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-26660-100ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Flow Cytometry, ELISA
Spécification
| CEACAM8/CD66b | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 24H12 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| A synthesized peptide derived from Human CEACAM8/CD66b [UniProt P31997] | |
| 100 μL | |
| Cancer | |
| 1088 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |