missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP290 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA536580
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 290 kDa. Purity is >95% by SDS-PAGE.
CEP290 Polyclonal antibody specifically detects CEP290 in Human, Mouse, Rat samples. It is validated for Western Blot.Spécification
CEP290 | |
Polyclonal | |
Unconjugated | |
Cep290 | |
3H11Ag; b2b1454Clo; b2b1752Clo; Bardet-Biedl syndrome 14 protein; Bardet-Biedl syndrome 14 protein homolog; BBS14; BC004690; Cancer/testis antigen 87; centrosomal protein 290; centrosomal protein 290kDa; centrosomal protein of 290 kDa; Cep290; CT87; CTCL tumor antigen se2-2; JBTS5; Kiaa0373; LCA10; Meckel syndrome, type 4; MKS4; monoclonal antibody 3H11 antigen; nephrocystin-6; nephrocytsin-6; Nphp6; POC3; POC3 centriolar protein homolog; prostate cancer antigen T21; rd16; RGD1311640; SLSN6; tumor antigen se2-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216274, 314787, 80184 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O15078, Q6A078 | |
Cep290 | |
Synthetic peptide corresponding to amino acids 781-830 of human CEP290 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |