missing translation for 'onlineSavingsMsg'
Learn More

CEP68 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15147-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834204

  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Centrosomal protein of 68 kDa is a protein that in humans is encoded by the CEP68 gene. It is mapped to chromosome 2. CEP68 is required for centrosome cohesion. It decorates fibres emanating from the proximal ends of centrioles. CEP68 and rootletin depend both on each other for centriole association, and both also require CEP250 for their function.
Spécification

Spécification

CEP68
Polyclonal
Unconjugated
CEP68
centrosomal protein 68kDa, Centrosomal protein of 68 kDa, CEP68, KIAA0582
Rabbit
Antigen Affinity Chromatography
RUO
23177
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q76N32
CEP68
CEP68 Fusion Protein Ag7447
20 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions