missing translation for 'onlineSavingsMsg'
Learn More

CEP89 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24002-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16842605

  • 140.00€ / 20µL
Expédition estimée: 30-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

24002-1-AP antibody recognizes all CEP123 isoforms.

CCDC123 (as known as CEP123), also named as CEP89, is a new player in the process of primary ciliogenesis and it also plays a role in mitochondrial metabolism where it may modulate complex IV activity. It has been shown that CEP123 is localized to the distal appendages of the mother centriole and the localization of CEP123 is cell cycle-dependent with its levels decreasing during mitosis. CEP123 depletion can cause defects in ciliary vesicle formation and prevent the formation of a ciliary vesicle at the distal end of the mother centriole. It is possible that CEP123 is involved in regulating the recruitment of membranes to the centrosome.
Spécification

Spécification

CEP89
Polyclonal
Unconjugated
CEP89
CCDC123, CEP123, CEP89
Rabbit
Antigen Affinity Chromatography
RUO
84902
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96ST8
CEP89
CEP89 Fusion Protein Ag21206
20 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions