missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP89 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24002-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
24002-1-AP antibody recognizes all CEP123 isoforms.
CCDC123 (as known as CEP123), also named as CEP89, is a new player in the process of primary ciliogenesis and it also plays a role in mitochondrial metabolism where it may modulate complex IV activity. It has been shown that CEP123 is localized to the distal appendages of the mother centriole and the localization of CEP123 is cell cycle-dependent with its levels decreasing during mitosis. CEP123 depletion can cause defects in ciliary vesicle formation and prevent the formation of a ciliary vesicle at the distal end of the mother centriole. It is possible that CEP123 is involved in regulating the recruitment of membranes to the centrosome.Spécification
CEP89 | |
Polyclonal | |
Unconjugated | |
CEP89 | |
CCDC123, CEP123, CEP89 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84902 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96ST8 | |
CEP89 | |
CEP89 Fusion Protein Ag21206 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |