Learn More
CHD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA565762
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Immunogen sequence: ALNLSVPRQRR RRRRKIEIEA ERAAKRRNLM EMVAQLRESQ VVSENGQEKV VDLSKASREA TSSTSNFSSL SSKFILPNVS TPVSDAFKTQ MEL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
This gene encodes a protein that contains several helicase family domains. Mutations in this gene have been found in some patients with the CHARGE syndrome.Spécification
CHD7 | |
Polyclonal | |
Unconjugated | |
CHD7 | |
A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55636 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2D1 | |
CHD7 | |
Recombinant Human CHD7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only