missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD7 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marque: Invitrogen PA547918
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
CHD7 Polyclonal antibody specifically detects CHD7 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spécification
CHD7 | |
Polyclonal | |
Unconjugated | |
CHD7 | |
A730019I05Rik; ATP-dependent helicase CHD7; CHARGE association; CHD7; CHD-7; chromodomain helicase DNA binding protein 7; chromodomain helicase DNA binding protein 7 isoform CRA_e; chromodomain-helicase-DNA-binding protein 7; CRG; Cycn; Cyn; Dz; Edy; Flo; GENA 47; Gena 52; GENA 60; HH5; IS3; KAL5; KIAA1416; Lda; metis; Mt; Obt; Todo; WBE1; Whi | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
55636 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9P2D1 | |
CHD7 | |
E. coli-derived recombinant human CHD7 Ala263-Gln457 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |