missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHIA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21484-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Degrades chitin and chitotriose. May participate in the defense against nematodes, fungi and other pathogens. Plays a role in T helper cell type 2 (Th2) immune response. Contributes to the response to IL-13 and inflammation in response to IL-13. Stimulates chemokine production by pulmonary epithelial cells. Protects lung epithelial cells against apoptosis and promotes phosphorylation of AKT1. Its function in the inflammatory response and in protecting cells against apoptosis is inhibited by allosamidin, suggesting that the function of this protein depends on carbohydrate binding.Spécification
CHIA | |
Polyclonal | |
Unconjugated | |
CHIA | |
2200003E03Rik, Acidic mammalian chitinase, AMCase, CHIA, CHIT2, chitinase, acidic, Lung specific protein TSA1902, RP5 1125M8.1, TSA1902 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113901, 27159, 81600 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6RY07, Q91XA9, Q9BZP6 | |
Chia | |
CHIA Fusion Protein Ag15767 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |