Learn More
Chordin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27293-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16835545
  • 399.00€ / 150µL
Expédition estimée: 30-05-2024
pour voir le stock disponible 
Description
Description
The Chordin antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Chordin. This antibody recognizes human, mouse antigen. The Chordin antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
Chordin
Polyclonal
Unconjugated
150 μL
8646
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human, Mouse