Learn More
Chordin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27293-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Chordin antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Chordin. This antibody recognizes human, mouse antigen. The Chordin antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Chordin | |
Polyclonal | |
Unconjugated | |
20 μL | |
8646 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |