Learn More
CHST4 Monoclonal Antibody (595223), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA524268
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human CHST1, 2, 5, 6, 10, 14, recombinant mouse CHST3, 4, and 7 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CHST4 Monoclonal antibody specifically detects CHST4 in Human samples. It is validated for Immunoprecipitation, Neutralization.Spécification
CHST4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8NCG5 | |
Chst4 | |
CHO-derived recombinant human Sulfotransferase 4/CHST4 His29-His386 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Neutralization | |
595223 | |
Unconjugated | |
Chst4 | |
carbohydrate (chondroitin 6/keratan) sulfotransferase 4; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; carbohydrate sulfotransferase 4; Chst4; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; galactose/N-acetylglucosamine/N-acetylglucosamine; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST2; GlcNAc6ST-2; Gn6st-2; Gst3; GST-3; HECGLCNAC6ST; HEC-GlcNAc6ST; high endothelial cell GlcNAC-6-sulphotransferase; high endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2; sulfo6 | |
Mouse | |
Protein A/G | |
RUO | |
10164 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |