Learn More
CLOCK Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human CLOCK.
Marque: Abnova PAB29318.100uL
Informations supplémentaires : Poids : 0.02000kg
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1. [provided by RefSeq]
Sequence: RQQEELRKIQEQLQMVHGQGLQMFLQQSNPGLNFGSVQLSSGNSSNIQQLAPINMQGQVVPTNQIQSGMNTGHIGTTQHMIQQQTLQSTSTQSQQNVLSGHSQQTSLPSQTQSTLTAPLYNTMVISQPAAGSMVQIPSSMPQNSTQSSpécification
CLOCK | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human CLOCK. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
CLOCK | |
Recombinant protein corresponding to amino acids of human CLOCK. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
KAT13D/KIAA0334/bHLHe8 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9575 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |