missing translation for 'onlineSavingsMsg'
Learn More

CLYBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17314-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837184

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CLYBL, also named as CLB, is a Mitochondrial citramalyl-CoA lyase indirectly involved in the vitamin B12 metabolism. It is a human mitochondrial enzyme of unknown function that is found in multiple eukaryotic taxa and conserved to bacteria. It acts as a beta-methylmalate synthase in vitro, by mediating conversion of glyoxylate and propionyl-CoA to beta-methylmalate. There are two isoforms with MW 31-33 kDa and 35-37 kDa.
Spécification

Spécification

CLYBL
Polyclonal
Unconjugated
CLYBL
citrate lyase beta like, CLB, CLYBL
Rabbit
Antigen Affinity Chromatography
RUO
171425, 306198, 69634
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I0K3, Q8N0X4, Q8R4N0
CLYBL
CLYBL Fusion Protein Ag11283
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions