missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24030-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
CMC1 | |
Polyclonal | |
Unconjugated | |
CMC1 | |
C3orf68, CMC1, Cmc1p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
152100, 67899 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7Z7K0, Q9CPZ8 | |
Cmc1 | |
CMC1 Fusion Protein Ag21224 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |