missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA524628
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
This antibody is predicted to react with bovine, chicken, porcine, non-human primate, rat and Xenopus based on sequence homology.
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.Spécification
Cofilin | |
Polyclonal | |
Unconjugated | |
CFL1 | |
18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1072, 12631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.25 mg/mL | |
PBS with 0.09% sodium azide | |
P18760, P23528 | |
CFL1 | |
KLH conjugated synthetic peptide between 70-98 amino acids from the central region of human CFL1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |