missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen Type VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14853-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The product of this gene contains several domains similar to von Willebrand Factor type A domains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in this gene are associated with Bethlem myopathy and Ullrich scleroatonic muscular dystrophy. Three transcript variants have been identified for this gene.Spécification
Collagen Type VI | |
Polyclonal | |
Unconjugated | |
COL6A2 | |
COL6A2, Collagen alpha 2(VI) chain, Collagen Type VI, collagen, type VI, alpha 2, PP3610 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12834, 1292, 361821 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12110, Q02788 | |
Col6a2 | |
Collagen Type VI Fusion Protein Ag6635 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |