Learn More
CORO2A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 10119-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16836293
  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
Description
The CORO2A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CORO2A. This antibody recognizes human, mouse antigen. The CORO2A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
CORO2A
Polyclonal
Unconjugated
150 μL
7464
IgG
ELISA, Immunohistochemistry, Western Blot
333 μg/mL
Rabbit
Primary
Mouse, Human