Learn More
CPAMD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA556705
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: YLPSYLSLGS WYSPSQCYLQ LQPPSHPLQV GEEAYFSVKS TCPCNFTLYY EVAARGNIVL SG Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 30%.
CPAMD8 Polyclonal antibody specifically detects CPAMD8 in Human samples. It is validated for Immunohistochemistry.Spécification
CPAMD8 | |
Polyclonal | |
Unconjugated | |
CPAMD8 | |
alpha-2 macroglobulin family protein VIP; C3 and PZP like, alpha-2-macroglobulin domain containing 8; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 8; C3 and PZP-like, alpha-2-macroglobulin domain containing 8; CPAMD8; K-CAP; VIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27151 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZJ3 | |
CPAMD8 | |
Recombinant protein corresponding to Human CPAMD8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only