missing translation for 'onlineSavingsMsg'
Learn More

CRMP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10525-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836963

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predomitly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
Spécification

Spécification

CRMP5
Polyclonal
Unconjugated
DPYSL5
CRAM, CRMP 5, CRMP3 associated molecule, CRMP5, dihydropyrimidinase like 5, DPYSL5, DRP 5, ULIP 6, Ulip6, UNC33 like phosphoprotein 6
Rabbit
Antigen Affinity Chromatography
RUO
56896, 65208, 65254
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BPU6, Q9EQF6, Q9JHU0
DPYSL5
CRMP5 Fusion Protein Ag0783
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions