missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRMP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10525-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predomitly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.Spécification
CRMP5 | |
Polyclonal | |
Unconjugated | |
DPYSL5 | |
CRAM, CRMP 5, CRMP3 associated molecule, CRMP5, dihydropyrimidinase like 5, DPYSL5, DRP 5, ULIP 6, Ulip6, UNC33 like phosphoprotein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56896, 65208, 65254 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BPU6, Q9EQF6, Q9JHU0 | |
DPYSL5 | |
CRMP5 Fusion Protein Ag0783 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |