Learn More
CSF2RB Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA548082
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human Common beta chain is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CSF2RB Polyclonal antibody specifically detects CSF2RB in Mouse samples. It is validated for Flow Cytometry, Neutralization, Western Blot.Spécification
CSF2RB | |
Polyclonal | |
Unconjugated | |
Csf2rb | |
Ai2ca; AI848964; AIC2A; Aic2b; Bc; beta c; beta common cytokine receptor; BetaIl3; Bil3; CD antigen CD131; CD131; CD131 antigen; CDw131; colony stimulating factor 2 receptor beta common subunit; colony stimulating factor 2 receptor, beta 1, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor 2 receptor subunit beta-2; colony-stimulating factor-2 receptor, beta, low-affinity; common beta chain; CSF2RB; Csf2rb1; Csf2rb2; Csfgmrb; cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL-3 receptor class 2 subunit beta; Il3r; IL3R beta; IL-3R class 2 subunit beta; IL3RB; Il3rb1; Il3rb2; IL5RB; interleukin 3 common beta chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12983, 12984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P26954, P26955 | |
Csf2rb, Csf2rb2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta His23-Trp440 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |