missing translation for 'onlineSavingsMsg'
Learn More

CST9L Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13649-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831974

  • 399.00€ / 150µL
Expédition estimée: 03-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The cystatin superfamily is a well-established family of cysteine protease inhibitors. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, while some also inhibit legumain family enzymes. CST9L (Cystatin-9-like) is a 147 amino acid secreted protein that is related to cystatin 9, which is thought to play a role in hematopoietic differentiation or inflammation. The gene encoding CST9L maps to human chromosome 20, which comprises approximately 2% of the human genome and contains nearly 63 million bases that encode over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought important for seminal production and may be potential targets for male contraception.
Spécification

Spécification

CST9L
Polyclonal
Unconjugated
CST9L
CST9L, cystatin 9 like
Rabbit
Antigen Affinity Chromatography
RUO
128821
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H4G1
CST9L
CST9L Fusion Protein Ag4084
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions