Learn More
CUL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 11107-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16827963
  • 121.38€ / 20µL
Expédition estimée: 29-09-2022
Description
This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.
Spécification
Spécification
CUL3
Polyclonal
Unconjugated
CUL3
CUL 3, CUL3, cullin 3, KIAA0617
Rabbit
Antigen Affinity Chromatography
RUO
26554, 301555, 8452
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B5DF89, Q13618, Q9JLV5
CUL3
CUL3 Fusion Protein Ag1555
20 μL
Primary
Human, Rat, Mouse