missing translation for 'onlineSavingsMsg'
Learn More

CXCL9/MIG Rabbit anti-Human, Clone: 50, Novus Biologicals™

Rabbit Monoclonal Antibody

Marque:  Novus Biologicals NBP2-89697-50UL

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 18253786

  • 392.00€ / 50µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY
Spécification

Spécification

CXCL9/MIG
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
50
ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Recombinant Human CXCL9/MIG protein
50 μg
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
4283
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.