missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXorf61 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25708-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CT83 is a protein coding gene.Spécification
CXorf61 | |
Polyclonal | |
Unconjugated | |
CT83 | |
Cancer/testis antigen 83, CT83, CXorf61, KK LC 1, KKLC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
203413 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5H943 | |
CT83 | |
CXorf61 Fusion Protein Ag22619 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |